(S)-1-[3-(Cyclopropyl-methyl-amino)-propyl]-1-(4-fluoro-phenyl)-1,3-dihydro-isobenzofuran-5-carbonitrile

Lieferant: Apollo Scientific

PC408411-500MG
APOSPC408411-500MGEA 865 EUR
APOSPC408411-500MG
(S)-1-[3-(Cyclopropyl-methyl-amino)-propyl]-1-(4-fluoro-phenyl)-1,3-dihydro-isobenzofuran-5-carbonitrile
(S)-1-[3-(Cyclopropyl-methyl-amino)-propyl]-1-(4-fluoro-phenyl)-1,3-dihydro-isobenzofuran-5-carbonitrile
Formel: C₂₂H₂₃FN₂O
Molecular Weight: 350,429 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24369843
CAS-Nummer: 1421026-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR