1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ylamine hydrochloride
Lieferant: Apollo Scientific
PC408412-500MG
APOSPC408412-500MGEA
865
EUR
APOSPC408412-500MG
1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ylamine hydrochloride
1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ylamine hydrochloride
Formel:
C₁₀H₁₁ClF₃N₃·ClH Molecular Weight: 302,124 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24369845 CAS-Nummer: 1420993-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...