4-(3-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC408413-500MG
APOSPC408413-500MGEA 786 EUR
APOSPC408413-500MG
4-(3-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
4-(3-Fluoro-benzenesulphinylmethyl)-piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₄FNO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD24369846
CAS-Nummer: 1420873-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR