2-[1-(2-Fluoro-benzyl)-piperidin-4-yl]-1H-benzoimidazole

Lieferant: Apollo Scientific

PC408414-500MG
APOSPC408414-500MGEA 865 EUR
APOSPC408414-500MG
2-[1-(2-Fluoro-benzyl)-piperidin-4-yl]-1H-benzoimidazole
2-[1-(2-Fluoro-benzyl)-piperidin-4-yl]-1H-benzoimidazole
Formel: C₁₉H₂₀FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD05254852
CAS-Nummer: 887217-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR