4-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidine-1-carboxylic acid tert-butyl ester
Lieferant: Apollo Scientific
Synonyme:
tert-Butyl 4-(1-(4-fluorobenzyl)-1H-benzo[d]imidazol-2-yl)piperidine-1-carboxylate
PC408415-500MG
APOSPC408415-500MGEA
865
EUR
APOSPC408415-500MG
4-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidine-1-carboxylic acid tert-butyl ester
4-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidine-1-carboxylic acid tert-butyl ester
Formel:
C₂₄H₂₈FN₃O₂ Molecular Weight: 409,496 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05662633 CAS-Nummer: 1420816-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...