1-(4-Fluoro-benzyl)-2-piperidin-4-yl-1H-benzoimidazole hydrochloride

Lieferant: Apollo Scientific

Synonyme: 1-(4-Fluorobenzyl)-2-(piperidin-4-yl)-1H-benzo[d]imidazole hydrochloride

PC408416-500MG
APOSPC408416-500MGEA 865 EUR
APOSPC408416-500MG
1-(4-Fluoro-benzyl)-2-piperidin-4-yl-1H-benzoimidazole hydrochloride
1-(4-Fluoro-benzyl)-2-piperidin-4-yl-1H-benzoimidazole hydrochloride
Formel: C₁₉H₂₀FN₃·ClH
Molecular Weight: 345,842 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24369855
CAS-Nummer: 1420975-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR