(3-Fluoro-phenyl)-methanesulphonic acid 1-(3-fluoro-phenylmethanesulphonyl)-piperidin-4-yl ester

Lieferant: Apollo Scientific

PC408417-500MG
APOSPC408417-500MGEA 619 EUR
APOSPC408417-500MG
(3-Fluoro-phenyl)-methanesulphonic acid 1-(3-fluoro-phenylmethanesulphonyl)-piperidin-4-yl ester
(3-Fluoro-phenyl)-methanesulphonic acid 1-(3-fluoro-phenylmethanesulphonyl)-piperidin-4-yl ester
Formel: C₁₉H₂₁F₂NO₅S₂
Molecular Weight: 445,501 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24369858
CAS-Nummer: 1420894-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR