(1-(5-Fluoropyrimidin-2-yl)piperidin-3-yl)methanamine hydrochloride

Lieferant: Apollo Scientific

PC408419-500MG
APOSPC408419-500MGEA 619 EUR
APOSPC408419-500MG
(1-(5-Fluoropyrimidin-2-yl)piperidin-3-yl)methanamine hydrochloride
(1-(5-Fluoropyrimidin-2-yl)piperidin-3-yl)methanamine hydrochloride
Formel: C₁₀H₁₅FN₄·ClH
Molecular Weight: 246,712 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22576794
CAS-Nummer: 1420880-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR