tert-Butyl ((1-(1-(4-fluorobenzyl)-1H-benzo[d]imidazol-2-yl)piperidin-4-yl)methyl)carbamate

Lieferant: Apollo Scientific

PC408420-500MG
APOSPC408420-500MGEA 619 EUR
APOSPC408420-500MG
tert-Butyl ((1-(1-(4-fluorobenzyl)-1H-benzo[d]imidazol-2-yl)piperidin-4-yl)methyl)carbamate
tert-Butyl ((1-(1-(4-fluorobenzyl)-1H-benzo[d]imidazol-2-yl)piperidin-4-yl)methyl)carbamate
Formel: C₂₅H₃₁FN₄O₂
Molecular Weight: 438,538 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24379041
CAS-Nummer: 1420888-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR