tert-Butyl 5-fluorospiro[indoline-3,4'-piperidine]-1'-carboxylate 96%

Lieferant: Apollo Scientific

PC408429-250MG
APOSPC408429-250MGEA 166 EUR
APOSPC408429-250MG
tert-Butyl 5-fluorospiro[indoline-3,4'-piperidine]-1'-carboxylate 96%
tert-Butyl 5-fluorospiro[indoline-3,4'-piperidine]-1'-carboxylate
Formel: C₁₇H₂₃FN₂O₂
Molecular Weight: 306,375 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198573
CAS-Nummer: 167484-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR