7-Fluorobenzo[b]thiophene-3-carboxylic acid 95%

Lieferant: Apollo Scientific

PC408448-100MG
APOSPC408448-100MGEA 705 EUR
APOSPC408448-100MG
7-Fluorobenzo[b]thiophene-3-carboxylic acid 95%
7-Fluorobenzo[b]thiophene-3-carboxylic acid
Formel: C₉H₅FO₂S
Molecular Weight: 196.198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28606191
CAS-Nummer: 1431309-15-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR