5-Bromo-6-fluoro-3-iodopyridin-2-amine 95%

Lieferant: Apollo Scientific

PC408449-100MG
APOSPC408449-100MGEA 429 EUR
APOSPC408449-100MG
5-Bromo-6-fluoro-3-iodopyridin-2-amine 95%
5-Bromo-6-fluoro-3-iodopyridin-2-amine
Formel: C₅H₃BrFIN₂
Molecular Weight: 316,898 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31631578
CAS-Nummer: 1207625-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR