4-(Difluoromethyl)piperidine hydrochloride 97%

Lieferant: Apollo Scientific

PC408512-250MG PC408512-1G PC408512-5G
APOSPC408512-250MGEA 133 EUR
APOSPC408512-250MG APOSPC408512-1G APOSPC408512-5G
4-(Difluoromethyl)piperidine hydrochloride 97%
4-(Difluoromethyl)piperidine hydrochloride
Formel: C₆H₁₁F₂N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD18711662
CAS-Nummer: 760958-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR