5-Trifluoromethyl-1H-indole-2-carboxylic acid 95+%

Lieferant: Apollo Scientific

Synonyme: 5-(Trifluoromethyl)-1H-indole-2-carboxylic acid

PC408513-250MG
APOSPC408513-250MGEA 139 EUR
APOSPC408513-250MG
5-Trifluoromethyl-1H-indole-2-carboxylic acid 95+%
5-Trifluoromethyl-1H-indole-2-carboxylic acid
Formel: C₁₀H₆F₃NO₂
Molecular Weight: 229,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095229
CAS-Nummer: 496946-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR