6-Fluoro-5-methyl-4-pyrimidinamine 95+%

Lieferant: Apollo Scientific

Synonyme: 6-Fluoro-5-methylpyrimidin-4-amine

PC408550-250MG
APOSPC408550-250MGEA 84.1 EUR
APOSPC408550-250MG
6-Fluoro-5-methyl-4-pyrimidinamine 95+%
6-Fluoro-5-methyl-4-pyrimidinamine
Formel: C₅H₆FN₃
Molecular Weight: 127.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10000602
CAS-Nummer: 18260-69-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR