4-(6-Chloro-2-trifluoromethylpyrimidin-4-yl)morpholine

Lieferant: Apollo Scientific

PC408551-1G
APOSPC408551-1GEA 177 EUR
APOSPC408551-1G
4-(6-Chloro-2-trifluoromethylpyrimidin-4-yl)morpholine
4-(6-Chloro-2-trifluoromethylpyrimidin-4-yl)morpholine
Formel: C₉H₉ClF₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD09743689
CAS-Nummer: 1189444-94-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR