4,6-Dichloro-5-fluoropyrimidin-2-amine

Lieferant: Apollo Scientific

PC408554-1G
APOSPC408554-1GEA 82.8 EUR
APOSPC408554-1G
4,6-Dichloro-5-fluoropyrimidin-2-amine
4,6-Dichloro-5-fluoropyrimidin-2-amine
Formel: C₄H₂Cl₂FN₃
Molecular Weight: 181.99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870221
CAS-Nummer: 15598-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR