5-Fluoro-2-methylpyrimidine-4,6-diol
Lieferant: Apollo Scientific
Synonyme:
5-Fluoro-6-hydroxy-2-methyl-4(1H)-pyrimidinone
PC408555-1G
APOSPC408555-1GEA
95.7
EUR
APOSPC408555-1G
5-Fluoro-2-methylpyrimidine-4,6-diol
5-Fluoro-2-methylpyrimidine-4,6-diol
Formel:
C₅H₅FN₂O₂ Molecular Weight: 144,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11520459 CAS-Nummer: 1598-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...