5-Fluoro-2-(methylsulphonyl)pyrimidine

Lieferant: Apollo Scientific

PC408558-1G
APOSPC408558-1GEA 184 EUR
APOSPC408558-1G
5-Fluoro-2-(methylsulphonyl)pyrimidine
5-Fluoro-2-(methylsulphonyl)pyrimidine
Formel: C₅H₅FN₂O₂S
Molecular Weight: 176,169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00223684
CAS-Nummer: 6090-38-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR