4-Chloro-5-fluoro-6-methylpyrimidine

Lieferant: Apollo Scientific

PC408559-1G
APOSPC408559-1GEA 454 EUR
APOSPC408559-1G
4-Chloro-5-fluoro-6-methylpyrimidine
4-Chloro-5-fluoro-6-methylpyrimidine
Formel: C₅H₄ClFN₂
Molecular Weight: 146,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16988365
CAS-Nummer: 898044-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR