2-Amino-4,6-difluorobenzoic acid

Lieferant: Apollo Scientific
Achtung

PC4086-250MG
APOSPC4086-250MGEA 21.9 EUR
APOSPC4086-250MG
2-Amino-4,6-difluorobenzoic acid
2-Amino-4,6-difluorobenzoic acid
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,1169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03618449
CAS-Nummer: 126674-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR