5-(Pentafluoroethyl)nicotinaldehyde

Lieferant: Apollo Scientific

PC408800-1G
APOSPC408800-1GEA 1510 EUR
APOSPC408800-1G
5-(Pentafluoroethyl)nicotinaldehyde
5-(Pentafluoroethyl)nicotinaldehyde
Formel: C₈H₄F₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD28122861
CAS-Nummer: 1816285-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR