2-Chloro-5-(pentafluoroethyl)pyridine

Lieferant: Apollo Scientific

PC408808-1G
APOSPC408808-1GEA 1870 EUR
APOSPC408808-1G
2-Chloro-5-(pentafluoroethyl)pyridine
2-Chloro-5-(pentafluoroethyl)pyridine
Formel: C₇H₃ClF₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD17677154
CAS-Nummer: 118559-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR