2-Cyano-6-(pentafluoroethyl)pyridine

Lieferant: Apollo Scientific

Synonyme: 2-Pyridinecarbonitrile, 6-(1,1,2,2,2-pentafluoroethyl)-, 6-Pentafluoroethylpyridine-2-carbonitrile, 6-(Pentafluoroethyl)-2-pyridinecarbonitrile

PC408814-1G
APOSPC408814-1GEA 1440 EUR
APOSPC408814-1G
2-Cyano-6-(pentafluoroethyl)pyridine
2-Cyano-6-(pentafluoroethyl)pyridine
Formel: C₈H₃F₅N₂
Molecular Weight: 222,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122873
CAS-Nummer: 1237431-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR