2-Cyano-5-(pentafluoroethyl)pyridine

Lieferant: Apollo Scientific

Synonyme: 5-(Pentafluoroethyl)-2-pyridinecarbonitrile, 2-Pyridinecarbonitrile, 5-(1,1,2,2,2-pentafluoroethyl)-, 5-(Pentafluoroethyl)pyridine-2-carbonitrile

PC408815-1G
APOSPC408815-1GEA 1440 EUR
APOSPC408815-1G
2-Cyano-5-(pentafluoroethyl)pyridine
2-Cyano-5-(pentafluoroethyl)pyridine
Formel: C₈H₃F₅N₂
Molecular Weight: 222,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122874
CAS-Nummer: 1421951-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR