2-(Pentafluoroethyl)benzonitrile

Lieferant: Apollo Scientific

PC408823-1G
APOSPC408823-1GEA 1440 EUR
APOSPC408823-1G
2-(Pentafluoroethyl)benzonitrile
2-(Pentafluoroethyl)benzonitrile
Formel: C₉H₄F₅N
Molecular Weight: 221,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122881
CAS-Nummer: 1823319-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR