3-(Pentafluoroethyl)benzonitrile

Lieferant: Apollo Scientific

PC408824-1G
APOSPC408824-1GEA 1440 EUR
APOSPC408824-1G
3-(Pentafluoroethyl)benzonitrile
3-(Pentafluoroethyl)benzonitrile
Formel: C₉H₄F₅N
Lagertemperatur: Raumtemperatur
MDL: MFCD28122882
CAS-Nummer: 1823246-09-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR