2-(Pentafluoroethyl)benzaldehyde

Lieferant: Apollo Scientific

PC408825-1G
APOSPC408825-1GEA 1730 EUR
APOSPC408825-1G
2-(Pentafluoroethyl)benzaldehyde
2-(Pentafluoroethyl)benzaldehyde
Formel: C₉H₅F₅O
Molecular Weight: 224,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22045188
CAS-Nummer: 133391-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR