Ethyl 4-(pentafluoroethyl)benzoate

Lieferant: Apollo Scientific

PC408828-1G
APOSPC408828-1GEA 1660 EUR
APOSPC408828-1G
Ethyl 4-(pentafluoroethyl)benzoate
Ethyl 4-(pentafluoroethyl)benzoate
Formel: C₁₁H₉F₅O₂
Molecular Weight: 268,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19439991
CAS-Nummer: 133512-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR