1-Nitro-4-(pentafluoroethyl)benzene

Lieferant: Apollo Scientific

PC408832-1G
APOSPC408832-1GEA 1510 EUR
APOSPC408832-1G
1-Nitro-4-(pentafluoroethyl)benzene
1-Nitro-4-(pentafluoroethyl)benzene
Formel: C₈H₄F₅NO₂
Molecular Weight: 241,115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122885
CAS-Nummer: 60979-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR