2-(Pentafluoroethyl)aniline
Lieferant: Apollo Scientific
PC408833-1G
APOSPC408833-1GEA
1440
EUR
APOSPC408833-1G
2-(Pentafluoroethyl)aniline
2-(Pentafluoroethyl)aniline
Formel:
C₈H₆F₅N Molecular Weight: 211,132 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24646210 CAS-Nummer: 60979-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...