3-(Pentafluoroethyl)aniline

Lieferant: Apollo Scientific

PC408834-1G
APOSPC408834-1GEA 1440 EUR
APOSPC408834-1G
3-(Pentafluoroethyl)aniline
3-(Pentafluoroethyl)aniline
Formel: C₈H₆F₅N
Molecular Weight: 211,132 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12196928
CAS-Nummer: 710-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR