2,6-Dichloro-3-(pentafluoroethyl)pyridine

Lieferant: Apollo Scientific

PC408838-1G
APOSPC408838-1GEA 1510 EUR
APOSPC408838-1G
2,6-Dichloro-3-(pentafluoroethyl)pyridine
2,6-Dichloro-3-(pentafluoroethyl)pyridine
Formel: C₇H₂Cl₂F₅N
Molecular Weight: 265,995 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122887
CAS-Nummer: 1816287-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR