2-Bromo-4-(pentafluoroethyl)pyridine
Lieferant: Apollo Scientific
PC408839-1G
APOSPC408839-1GEA
2020
EUR
APOSPC408839-1G
2-Bromo-4-(pentafluoroethyl)pyridine
2-Bromo-4-(pentafluoroethyl)pyridine
Formel:
C₇H₃BrF₅N Molecular Weight: 276,001 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28122888 CAS-Nummer: 1816286-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...