3-Bromo-4-(pentafluoroethyl)pyridine
Lieferant: Apollo Scientific
PC408840-1G
APOSPC408840-1GEA
2310
EUR
APOSPC408840-1G
3-Bromo-4-(pentafluoroethyl)pyridine
3-Bromo-4-(pentafluoroethyl)pyridine
Formel:
C₇H₃BrF₅N Molecular Weight: 276,001 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28122889 CAS-Nummer: 1816286-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...