3-Bromo-2-(pentafluoroethyl)pyridine

Lieferant: Apollo Scientific

PC408841-1G
APOSPC408841-1GEA 1730 EUR
APOSPC408841-1G
3-Bromo-2-(pentafluoroethyl)pyridine
3-Bromo-2-(pentafluoroethyl)pyridine
Formel: C₇H₃BrF₅N
Molecular Weight: 276,001 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122890
CAS-Nummer: 1816283-70-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR