2-(Pentafluoroethyl)isonicotinaldehyde

Lieferant: Apollo Scientific

PC408846-1G
APOSPC408846-1GEA 2310 EUR
APOSPC408846-1G
2-(Pentafluoroethyl)isonicotinaldehyde
2-(Pentafluoroethyl)isonicotinaldehyde
Formel: C₈H₄F₅NO
Lagertemperatur: Raumtemperatur
MDL: MFCD28122894
CAS-Nummer: 1816283-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR