6-(Pentafluoroethyl)pyridine-3-carbaldehyde

Lieferant: Apollo Scientific

PC408847-1G
APOSPC408847-1GEA 1510 EUR
APOSPC408847-1G
6-(Pentafluoroethyl)pyridine-3-carbaldehyde
6-(Pentafluoroethyl)pyridine-3-carbaldehyde
Formel: C₈H₄F₅NO
Molecular Weight: 225,115 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28122895
CAS-Nummer: 1816282-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR