N-t-BOC-cis-4-Fluoro-L-proline methyl ester 95+%

Lieferant: Apollo Scientific

Synonyme: Boc-cis-4-F-Pro-OMe, (2S,4S)-1-tert-Butyl 2-methyl 4-fluoropyrrolidine-1,2-dicarboxylate

PC409006-1G
APOSPC409006-1GEA 119 EUR
APOSPC409006-1G
N-t-BOC-cis-4-Fluoro-L-proline methyl ester 95+%
N-t-BOC-cis-4-Fluoro-L-proline methyl ester
Formel: C₁₁H₁₈FNO₄
Molecular Weight: 247.263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054130
CAS-Nummer: 203866-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR