1-Boc-3-fluoro-4-pyrrolidinone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate, 4-Fluoropyrrolidin-3-one, N-BOC protected
PC409012-1G
APOSPC409012-1GEA
394
EUR
APOSPC409012-1G
1-Boc-3-fluoro-4-pyrrolidinone
1-Boc-3-fluoro-4-pyrrolidinone
Formel:
C₉H₁₄FNO₃ Molecular Weight: 203,213 g/mol Schmelzpunkt: 81…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08274459 CAS-Nummer: 845894-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...