N-BOC-cis-4-Trifluoromethyl-L-proline

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S,4S)-1-(tert-Butoxycarbonyl)-4-(trifluoromethyl)pyrrolidine-2-carboxylic acid, (4S)-1-(tert-Butoxycarbonyl)-4-(trifluoromethyl)-L-proline

PC409016-1G
APOSPC409016-1GEA 1090 EUR
APOSPC409016-1G
N-BOC-cis-4-Trifluoromethyl-L-proline
N-BOC-cis-4-Trifluoromethyl-L-proline
Formel: C₁₁H₁₆F₃NO₄
Molecular Weight: 283,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08460520
CAS-Nummer: 470482-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR