3,3-Difluorocyclobutyl benzoate

Lieferant: Apollo Scientific

PC409026-100MG
APOSPC409026-100MGEA 836 EUR
APOSPC409026-100MG
3,3-Difluorocyclobutyl benzoate
3,3-Difluorocyclobutyl benzoate
Formel: C₁₁H₁₀F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16039588
CAS-Nummer: 1215071-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR