4-Fluorocyclohexanone

Lieferant: Apollo Scientific
Achtung

PC409038-1G PC409038-250MG
APOSPC409038-1GEA 1060 EUR
APOSPC409038-1G APOSPC409038-250MG
4-Fluorocyclohexanone
4-Fluorocyclohexanone
Formel: C₆H₉FO
Lagertemperatur: Raumtemperatur
MDL: MFCD11520472
CAS-Nummer: 68223-64-3
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR