(3,3-Difluorocyclobutyl)ethanol

Lieferant: Apollo Scientific
Achtung

PC409042-1G
APOSPC409042-1GEA 1390 EUR
APOSPC409042-1G
(3,3-Difluorocyclobutyl)ethanol
(3,3-Difluorocyclobutyl)ethanol
Formel: C₆H₁₀F₂O
Molecular Weight: 136.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18790781
CAS-Nummer: 1056467-54-9
UN: 1987
ADR: 3 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR