Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate 95+%

Lieferant: Apollo Scientific

PC409044-250MG
APOSPC409044-250MGEA 104 EUR
APOSPC409044-250MG
Ethyl-1-benzyl-5,5-difluoro-4-oxo-piperidine-3-carboxylate 95+%
Formel: C₁₅H₁₇F₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16036533
CAS-Nummer: 1067915-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR