(3,3-Difluorocyclobutyl)acetic acid

Lieferant: Apollo Scientific
Achtung

PC409045-250MG
APOSPC409045-250MGEA 311 EUR
APOSPC409045-250MG
(3,3-Difluorocyclobutyl)acetic acid
(3,3-Difluorocyclobutyl)acetic acid
Formel: C₆H₈F₂O₂
Molecular Weight: 150,123 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22199410
CAS-Nummer: 1373503-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR