N-t-BOC-5,5-Difluoropiperidine-3-Methanol

Lieferant: Apollo Scientific

PC409052-250MG
APOSPC409052-250MGEA 1060 EUR
APOSPC409052-250MG
N-t-BOC-5,5-Difluoropiperidine-3-Methanol
N-t-BOC-5,5-Difluoropiperidine-3-Methanol
Formel: C₁₁H₁₉F₂NO₃
Molecular Weight: 251,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16875553
CAS-Nummer: 1262412-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR