4-Fluoro-4-methylpiperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

PC409056-250MG
APOSPC409056-250MGEA 676 EUR
APOSPC409056-250MG
4-Fluoro-4-methylpiperidine hydrochloride
4-Fluoro-4-methylpiperidine hydrochloride
Formel: C₆H₁₂FN·ClH
Molecular Weight: 153,626 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381135
CAS-Nummer: 1023305-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR