3-(2-Fluorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%

Lieferant: Apollo Scientific

PC409514-1G
APOSPC409514-1GEA 957 EUR
APOSPC409514-1G
3-(2-Fluorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine 95+%
3-(2-Fluorophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine
Formel: C₁₂H₁₂FN₃
Molecular Weight: 217.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258791
CAS-Nummer: 916423-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR