2,2,2-Trifluoro-1-(6-(trifluoromethyl)pyridin-3-yl)ethanol 95+%

Lieferant: Apollo Scientific

PC409525-1G
APOSPC409525-1GEA 365 EUR
APOSPC409525-1G
2,2,2-Trifluoro-1-(6-(trifluoromethyl)pyridin-3-yl)ethanol 95+%
2,2,2-Trifluoro-1-(6-(trifluoromethyl)pyridin-3-yl)ethanol
Formel: C₈H₅F₆NO
Lagertemperatur: Raumtemperatur
MDL: MFCD11226776
CAS-Nummer: 1092353-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR